Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropeptide FF R1/NPFFR1 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NB300168

 View more versions of this product

Catalog No. NB300168


Only null left
Add to Cart

Description

Description

Neuropeptide FF R1/NPFFR1 Polyclonal specifically detects Neuropeptide FF R1/NPFFR1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Neuropeptide FF R1/NPFFR1
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
NPFFR1
N terminal sequence MEGEPSQPPNSSWPLS and the C terminal sequence CSHLPLTIPAWDI of the human NPFF1 protein.
0.1 mg
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
64106
Human
IgG
Western Blot
1 mg/mL
Western Blot 1:500 -1:1000
FLJ10751, G protein-coupled receptor 147, GPR147, neuropeptide FF 1, neuropeptide FF receptor 1, NPFF1G-protein coupled receptor 147, NPFF1R1, OT7T022, RFamide-related peptide receptor OT7T022
Rabbit
Affinity Purified
RUO
Primary
Reacts with the N terminal sequence MEGEPSQPPNSSWPLS and the C terminal sequence CSHLPLTIPAWDI of the human NPFF1 protein.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only