Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropeptide FF R1/NPFFR1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB362230 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362230 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362230 Supplier Novus Biologicals Supplier No. NB300168MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Neuropeptide FF R1/NPFFR1 Polyclonal antibody specifically detects Neuropeptide FF R1/NPFFR1 in Human samples. It is validated for Western Blot

Specifications

Antigen Neuropeptide FF R1/NPFFR1
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FLJ10751, G protein-coupled receptor 147, GPR147, neuropeptide FF 1, neuropeptide FF receptor 1, NPFF1G-protein coupled receptor 147, NPFF1R1, OT7T022, RFamide-related peptide receptor OT7T022
Host Species Rabbit
Immunogen N terminal sequence MEGEPSQPPNSSWPLS and the C terminal sequence CSHLPLTIPAWDI of the human NPFF1 protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 64106
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.