Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropeptide Y Antibody (8) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP105166SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105166SS 0.025 mg
NBP105166 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP105166SS Supplier Novus Biologicals Supplier No. NBP105166SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neuropeptide Y Monoclonal specifically detects Neuropeptide Y in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Neuropeptide Y
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:16000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide
Gene Alias 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase
Gene Symbols NPY
Host Species Mouse
Immunogen Synthetic human/rat NPY coupled to carrier with glutaraldehyde
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline GPCR, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 4852
Test Specificity Binds full length human/rat NPY. Cross-reactivity with NPY (1-24), NPY (18-36), PYY or PP was <0.2%.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.