Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuropeptide Y Antibody (8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $560.50
Specifications
Antigen | Neuropeptide Y |
---|---|
Clone | 8 |
Concentration | 1.0 mg/mL |
Dilution | ELISA 1:16000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP105166SS
![]() |
Novus Biologicals
NBP105166SS |
0.025 mg |
Each for $268.00
|
|
|||||
NBP105166
![]() |
Novus Biologicals
NBP105166 |
0.1 mg |
Each for $560.50
|
|
|||||
Description
Neuropeptide Y Monoclonal specifically detects Neuropeptide Y in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Neuropeptide Y | |
1.0 mg/mL | |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
GPCR, Neuroscience, Neurotransmission | |
170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase | |
NPY | |
IgG1 κ | |
Protein A or G purified | |
Binds full length human/rat NPY. Cross-reactivity with NPY (1-24), NPY (18-36), PYY or PP was <0.2%. |
8 | |
ELISA 1:16000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
4852 | |
Synthetic human/rat NPY coupled to carrier with glutaraldehyde | |
Primary | |
Store at 4C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title