Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropeptide Y Antibody, Novus Biologicals™
SDP

Catalog No. NBP146535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.15 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146535 0.15 mL
NBP146535SS 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP146535 Supplier Novus Biologicals Supplier No. NBP146535
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Neuropeptide Y Polyclonal antibody specifically detects Neuropeptide Y in Human, Mouse, Rat, Rabbit, Xenopus species samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry - Free Floating.

Specifications

Antigen Neuropeptide Y
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:300-1:3000, Immunohistochemistry 1:300-1:3000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:300-1:3000, Immunohistochemistry-Frozen 1:300-1:3000, Immunohistochemistry Free-Floating
Formulation Lyophilized from whole antisera with No Preservative
Gene Accession No. P57774
Gene Alias 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase
Gene Symbols NPY
Host Species Goat
Immunogen A synthetic peptide from amino acid region 20-70 of mouse Neuropeptide Y conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in many other species including human, rat, zebrafish and Xenopus species.
Purification Method Unpurified
Quantity 0.15 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 4852
Test Specificity Specific for NPY.
Reconstitution Reconstitute in 150 ul of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Target Species Human, Mouse, Rat, Rabbit, Xenopus species, Zebrafish
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.