Learn More
Description
Specifications
Specifications
| Antigen | Neuropeptide Y |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:300-1:3000, Immunohistochemistry 1:300-1:3000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:300-1:3000, Immunohistochemistry-Frozen 1:300-1:3000, Immunohistochemistry Free-Floating |
| Formulation | Lyophilized from whole antisera with No Preservative |
| Gene Accession No. | P57774 |
| Gene Alias | 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase |
| Gene Symbols | NPY |
| Host Species | Goat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
