Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropilin-1 Antibody (130603), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB303238 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303238 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303238 Supplier Novus Biologicals Supplier No. FAB566X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neuropilin-1 Monoclonal specifically detects Neuropilin-1 in Rat samples. It is validated for Blockade of Receptor-ligand Interaction, CyTOF, Flow Cytometry.

Specifications

Antigen Neuropilin-1
Applications Blockade of Receptor-ligand Interaction, CyTOF, Flow Cytometry
Classification Monoclonal
Clone 130603
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias BDCA4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, NP1, NRP, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-1, Phe22-Asp854 (Lys811Arg, Pro812-Gly828 del), predicted, Accession # Q9QWJ9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Breast Cancer, Cancer, Cardiovascular Biology, Neuroscience, Phospho Specific, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 8829
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.