Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropilin-1 Antibody (211H6.01), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. DDX0440A488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0440A488 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0440A488 Supplier Novus Biologicals Supplier No. DDX0440A488100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neuropilin-1 Monoclonal specifically detects Neuropilin-1 in Canine, Human samples. It is validated for Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Neuropilin-1
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 211H6.01
Concentration 0.5mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry
Gene Accession No. NP_003864
Gene Alias BDCA4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, NP1, NRP, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R
Gene Symbols NRP1
Host Species Mouse
Immunogen Neuropilin-1 Antibody (211H6.01) was made to Neuropilin/CD304.
Molecular Weight of Antigen 103 kDa
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Breast Cancer, Cancer, Cardiovascular Biology, Neuroscience, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 8829
Target Species Canine, Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.