Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropilin-1 Antibody (446921), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB371321 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371321 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371321 Supplier Novus Biologicals Supplier No. FAB3870Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neuropilin-1 Monoclonal specifically detects Neuropilin-1 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Neuropilin-1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 446921
Conjugate DyLight 680
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias BDCA4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, NP1, NRP, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Neuropilin-1, Phe22-Lys644, Accession # NP_001019799
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Breast Cancer, Cancer, Cardiovascular Biology, Neuroscience, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 8829
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.