Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropilin-2 Antibody (257103), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB349173 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349173 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349173 Supplier Novus Biologicals Supplier No. FAB22151MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neuropilin-2 Monoclonal specifically detects Neuropilin-2 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Neuropilin-2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 257103
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias MGC126574, neuropilin 2, neuropilin-2, neuropilin-2a(22), neuropilin-2b(0), NP2, NPN 2, NPN2, PRO2714, receptor for VEGF165 and semaphorins class3, Vascular endothelial cell growth factor 165 receptor 2, vascular endothelial growth factor-165 receptor 2, VEGF1265R2, VEGF165R2neuropilin-2a(17)
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Neuropilin-2, Gln23-Tyr855, Accession # NP_003863
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 8828
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.