Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropilin-2 Antibody (96009R) [HRP], Novus Biologicals™
SDP

Catalog No. NB377700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377700 Supplier Novus Biologicals Supplier No. FAB567RH
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neuropilin-2 Monoclonal specifically detects Neuropilin-2 in Rat samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen Neuropilin-2
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 96009R
Conjugate HRP
Dilution Western Blot, Simple Western, Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias MGC126574, neuropilin 2, neuropilin-2, neuropilin-2a(22), neuropilin-2b(0), NP2, NPN 2, NPN2, PRO2714, receptor for VEGF165 and semaphorins class3, Vascular endothelial cell growth factor 165 receptor 2, vascular endothelial growth factor-165 receptor 2, VEGF1265R2, VEGF165R2neuropilin-2a(17)
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-2, Gln23-Asp857 (Val809-Asp825 del), Accession # O35276
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 8828
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.