Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuroplastin Antibody (OTI3C4), FITC, Novus Biologicals™
SDP

Catalog No. NBP272966F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272966F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272966F Supplier Novus Biologicals Supplier No. NBP272966F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neuroplastin Monoclonal specifically detects Neuroplastin in Human, Rat, Canine, Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Neuroplastin
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3C4
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias GP55Stromal cell-derived receptor 1, GP65SDR-1, MGC102805, neuroplastin, np55, np65, SDFR1DKFZp686L2477, SDR1stromal cell derived factor receptor 1
Gene Symbols NPTN
Host Species Mouse
Immunogen Full length recombinant protein of human Neuroplastin (NP_059429) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 27020.0
Target Species Human, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.