Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutrophil Elastase/ELA2 Antibody (39A) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017572 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017572 0.025 mg
NB10064392 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017572 Supplier Novus Biologicals Supplier No. NB100643920.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Elastase Monoclonal specifically detects Elastase in Human samples. It is validated for ELISA, Flow Cytometry, Functional.

Specifications

Antigen Elastase
Applications ELISA, Flow Cytometry, Functional Assay
Classification Monoclonal
Clone 39A
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Functional
Formulation PBS (pH 7.2) with 0.09% Sodium Azide
Gene Accession No. P08246
Gene Alias CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E
Gene Symbols ELANE
Host Species Mouse
Immunogen Elastase from human neutrophils
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23436
Test Specificity NB100-64392 is specific for human neutrophil elastase.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.