Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neutrophil Elastase/ELA2 Antibody (39A) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$206.50 - $499.50
Specifications
Antigen | Elastase |
---|---|
Clone | 39A |
Concentration | 0.5 mg/mL |
Dilution | Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Functional |
Applications | Functional Assay, Flow Cytometry, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017572
![]() |
Novus Biologicals
NB100643920.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10064392
![]() |
Novus Biologicals
NB10064392 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
Elastase | |
0.5 mg/mL | |
Functional Assay, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
Human | |
CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E | |
ELANE | |
IgG1 | |
Protein G purified | |
NB100-64392 is specific for human neutrophil elastase. |
39A | |
Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Functional | |
Monoclonal | |
Purified | |
RUO | |
P08246 | |
23436 | |
Elastase from human neutrophils | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title