Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutrophil Elastase/ELA2 Antibody (39A), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB10064392C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064392C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064392C Supplier Novus Biologicals Supplier No. NB10064392C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for ELISA, Flow Cytometry.

Specifications

Antigen Neutrophil Elastase/ELA2
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone 39A
Conjugate DyLight 650
Dilution Flow Cytometry, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1
Gene Symbols ELANE
Host Species Mouse
Immunogen Elastase from human neutrophils
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1991.0
Test Specificity NB100-64392 is specific for human neutrophil elastase.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.