Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutrophil Elastase/ELA2 Antibody (950312), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB302456 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302456 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302456 Supplier Novus Biologicals Supplier No. FAB91673G

Mouse Monoclonal Antibody

Neutrophil Elastase/ELA2 Monoclonal antibody specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for ELISA, Flow Cytometry, CyTOF

Specifications

Antigen Neutrophil Elastase/ELA2
Applications ELISA, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 950312
Conjugate Alexa Fluor 488
Dilution ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Elastase/ELA-2, Met1-Asn252, Accession # P08246
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1991
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.