Learn More
Neutrophil Elastase/ELA2 Antibody (950334) - Azide and BSA Free, R&D Systems™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Neutrophil Elastase/ELA2 |
| Applications | CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 950334 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready |
| Formulation | PBS with No Preservative |
| Gene Alias | Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 |
| Gene Symbols | ELANE |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.