Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutrophil Elastase/ELA2 Antibody (NP57), HRP, Novus Biologicals™
SDP

Catalog No. NBP250529H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250529H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250529H Supplier Novus Biologicals Supplier No. NBP250529H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Neutrophil Elastase/ELA2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone NP57
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1
Gene Symbols ELANE
Host Species Mouse
Immunogen Neutrophil granule proteins
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1991
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.