Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutrophil Elastase/ELA2 Mouse anti-Human, Clone: AHN-10, Novus Biologicals™
SDP

Catalog No. NBP229778 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229778 0.1 mL
NB019461 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP229778 Supplier Novus Biologicals Supplier No. NBP229778

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Radioimmunoassay.

Specifications

Antigen Neutrophil Elastase/ELA2
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone AHN-10
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Radioimmunoassay 1:100-1:2000
Formulation Ascites with No Preservative
Gene Accession No. P08246
Gene Alias Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1
Gene Symbols ELANE
Host Species Mouse
Immunogen This antibody is specific to Neutrophil Elastase
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1991
Test Specificity Reacts specifically with human neutrophil elastase and not with the 3 homologous neutrophil neutral serine proteases, cathepsin G, esterase N and proteinase 3. Inhibits the elastinolytic activity of elastase. Elastase is felt to play an important role in a number of disease states.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.