Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutrophils Antibody (Nimp-R14), PerCP, Novus Biologicals™
SDP

Catalog No. NB173036 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173036 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173036 Supplier Novus Biologicals Supplier No. NBP280078PCP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Neutrophils Monoclonal antibody specifically detects Neutrophils in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen Neutrophils
Applications Flow Cytometry
Classification Monoclonal
Clone Nimp-R14
Conjugate PerCP
Formulation PBS
Host Species Rat
Immunogen Purified mouse BALB/c neutrophils.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.