Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutrophils Antibody (RM0028-3G23) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB019220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019220 0.025 mg
NBP2124391 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019220 Supplier Novus Biologicals Supplier No. NBP2124390.025MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Neutrophils Monoclonal specifically detects Neutrophils in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Neutrophils
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone RM0028-3G23
Conjugate Unconjugated
Dilution Western Blot 1:500-1000, Immunohistochemistry 1:100-300, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:100-300
Formulation A 0.2 um filtered solution in PBS with No Preservative
Host Species Rat
Immunogen Isolated mouse neutrophil cells
Purification Method Protein G purified
Quantity 0.025 mg
Primary or Secondary Primary
Reconstitution Reconstitute with 0.4 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Mouse
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.