Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Newcastle Disease Virus (NDV) Antibody (9F7), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB359817 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359817 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359817 Supplier Novus Biologicals Supplier No. NB11059912JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Newcastle Disease Virus (NDV) Monoclonal antibody specifically detects Newcastle Disease Virus (NDV) in Virus samples. It is validated for Western Blot, Haemagglutination, ELISA

Specifications

Antigen Newcastle Disease Virus (NDV)
Applications Western Blot, Hemagglutination Assay, Sandwich ELISA
Classification Monoclonal
Clone 9F7
Conjugate Janelia Fluor 646
Dilution Western Blot, Haemagglutination, Sandwich ELISA
Formulation 50mM Sodium Borate
Gene Alias NDV
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Newcastle disease virus (La-sota strain).
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.