Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Newtex™ ZetexPlus™ 200 Series Heat Resistant Mitts
Shop All Newtex Industries, Inc. Products

Click to view available options
Length (English):
11 in.
23 in.
Description
- Vermiculite coating evenly dissipates heat across the surface of the glove
- Coating significantly increases temperature rating and abrasion resistance
- Premium wool liner
- Double stitched and reinforced
- Inorganic fibers will not produce hazardous outgassing
Specifications
Specifications
Coating Material | Vermiculite |
Color | Tan |
Hand | Ambidextrous |
Material | ZetexPlus Fiberglass Fabric |
Size | Universal |
Temperature (English) Max. | 1500°F (Extended), 2000°F (Intermittent) |
Temperature (Metric) Max | 815°C (Extended), 1095°C (Intermittent) |
For Use With (Application) | Kiln/Furnace Operations, Heat Processing Facilities, and High Temperature Industrial Environments |
Glove Cuff Style | Gauntlet |
Liner Material | Wool |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction