Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nexilin Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31740225UL
Description
Nexilin Polyclonal antibody specifically detects Nexilin in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Nexilin | |
Polyclonal | |
Western Blot 0.04 to 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
CMD1CC, F-actin-binding protein, MGC104234, MGC138865, MGC138866, Nelin, NELINnexilin, nexilin (F actin binding protein) | |
This antibody was developed against Recombinant Protein corresponding to amino acids: EEERARRRAIDLEIKEREAENFHEEDDVDVRPARKSEAPFTHKVNMKARFEQMAKAREEEEQRRIEEQKLLRMQFEQREIDAALQKKREEEEEEEGSIMNGSTAEDEEQTRSGA | |
25 μg | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
91624 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction