Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nexilin Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$343.50 - $573.00

Specifications

Antigen Nexilin
Dilution Western Blot 0.04 to 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB169880
SDP
View Documents
Novus Biologicals
NBP31740225UL
25 μg
Each for $343.50
Only null left
Add to Cart
 
NB169879
SDP
View Documents
Novus Biologicals
NBP317402100UL
100 μg
Each for $573.00
Only null left
Add to Cart
 
Description

Description

Nexilin Polyclonal antibody specifically detects Nexilin in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

Nexilin
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Cell Biology, Cytoskeleton Markers, Signal Transduction
PBS, pH 7.2, 40% glycerol
91624
IgG
Affinity purified
Western Blot 0.04 to 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Polyclonal
Purified
RUO
Human
CMD1CC, F-actin-binding protein, MGC104234, MGC138865, MGC138866, Nelin, NELINnexilin, nexilin (F actin binding protein)
This antibody was developed against Recombinant Protein corresponding to amino acids: EEERARRRAIDLEIKEREAENFHEEDDVDVRPARKSEAPFTHKVNMKARFEQMAKAREEEEQRRIEEQKLLRMQFEQREIDAALQKKREEEEEEEGSIMNGSTAEDEEQTRSGA
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.