Learn More
Nextteq™ Nextteq™ Gastec™ Detector Tubes: : Acetic Acid
SDP
Catalog No. 19026345 Shop All Nextteq Products
Change view
Click to view available options
Measurement Range:
0.125 to 25
1 to 100
Response Time:
1.5 min.
30 sec. to 1.5 min.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Response Time
19-026-345 1 to 100 30 sec. to 1.5 min.
19-026-348 0.125 to 25 1.5 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-026-345 Supplier Nextteq™ Supplier No. 81
Add to Cart
Add to Cart
This item is not returnable. View return policy
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.
Specifications
Components Measured Acetic Acid
Measurement Range 1 to 100
No. of Strokes 0.5 to 2
Quantity 10
Response Time 30 sec. to 1.5 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C