Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nextteq™ Nextteq™ Gastec™ Detector Tubes: : Acetic Acid
SDP

Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.

$172.00 - $267.50

Specifications

Components Measured Acetic Acid
No. of Strokes 0.5 to 2
Quantity 10
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
View More Specs

Products 2
Catalog Number Mfr. No. Components Measured Measurement Range Response Time No. of Strokes Price Quantity & Availability  
Catalog Number Mfr. No. Components Measured Measurement Range Response Time No. of Strokes Price Quantity & Availability  
19-026-348
SDP
View Documents
Nextteq™
81L
Acetic Acid 0.125 to 25 1.5 min. 0.5 to 2
Pack of 10 for $172.00
Case of 10 PK for $1,496.00
Only null left
Add to Cart
 
19-026-345
SDP
View Documents
Nextteq™
81
Acetic Acid 1 to 100 30 sec. to 1.5 min. 0.5 to 2
Pack of 10 for $267.50
Only null left
Add to Cart
 
Description

Description

Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.
Specifications

Specifications

Acetic Acid
10
0°C to 40°C
0.5 to 2
32°F to 104°F
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.