Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF-1A Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$156.65 - $620.58
Specifications
Antigen | NF-1A |
---|---|
Concentration | Conc. Not Determined |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-4027
![]() ![]() |
Active Motif
39398 |
10 μL |
Each for $156.65
|
|
|||||
50-199-4026
![]() ![]() |
Active Motif
39397 |
100 μL |
Each for $620.58
|
|
|||||
Description
NF-1, also designated CTF, consists of a family of CCAAT box binding proteins that stimulate DNA replication and activate transcription. Analysis of human NF-1 messenger RNA has revealed two forms of the NF-1 protein arising from an alternate splicing of a single NF-1 gene. NF-1 binds its consensus DNA element as a homodimer via an amino-terminal DNA binding domain, and activates transcription through a putatively novel, proline-rich, carboxy terminal transactivation domain. The NF-1 protein has been shown to recognize and bind the adenovirus type 2 promoter and activate transcription of herpes simplex virus thymidine kinase genes. The NF-1 consensus element has been found in the upstream promoter region of myriad eukaryotic genes, including that of Ha-Ras, alpha-globin, HSP 70, GRP 78, Histone H1, myelin basic protein and in the Xenopus laevis vitellogenin gene promoter.Specifications
NF-1A | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q14938 | |
4784 | |
This NF-1A antibody was raised against a synthetic peptide corresponding to amino acid residue(s) 478-492 of human NF-1A. | |
Primary | |
Antibody |
Conc. Not Determined | |
Polyclonal | |
Liquid | |
RUO | |
whole serum with 30% glycerol and 0.035% sodium azide; pH | |
CCAAT-binding transcription factor; CCAAT-box-binding transcription factor; CTF; KIAA1439; MRSHSS; NF1A; NF1-A; NF1-B; NF1-C; NF1-X; NF1-X1 protein; NFI; NF-I/A; NF-I/B; NF-I/C; NF-I/X; NFIA; NFI-A; NFIB; NFI-B; NFIC; NFI-C; Nfix; NFI-X; Nuclear factor 1 X-type; Nuclear factor 1/A; Nuclear factor 1/B; Nuclear factor 1/C; nuclear factor 1/X; nuclear factor I X; Nuclear factor I/A; Nuclear factor I/B; Nuclear factor I/C; Nuclear factor I/X; nuclear factor I/X (CCAAT-binding transcription factor); SOTOS2; TGGCA-binding protein | |
NFIX | |
IgG | |
-20°C, Avoid Freeze/Thaw Cycles | |
NFIX |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title