Learn More
Description
This product is supplied with a high degree of hydration and some residual NaCl, the amount of which are batch dependent. Please refer to the Certificate of Analysis to obtain the batch specific Net Product Content and the maximum solubility threshold to use in dilution calculations.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 1505.06 |
| Purity | 90% |
| Molecular Weight (g/mol) | 1439.109 |
| Chemical Name or Material | NF 449 |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

