Learn More
CellSensor™ CellSensor™ NFκB-bla THP-1 Cell Line
Description
- The CellSensor™ NFκB-bla THP-1 cell line contains a beta-lactamase reporter gene under the control of the Nuclear Factor kappa B (NFκB) response element stably integrated into THP-1 cells.
- To obtain the cell line, the NFκB-bla construct was transduced into THP-1 cells by lentivirus.
- Subsequent flow cytometry was used to isolate cells responsive to Tumor Necrosis Factor alpha (TNFα).
- The cell line was validated for DMSO tolerance, cell number, stimulation time, substrate loading time, and EC50 concentration of TNFα.
- The CellSensor™ NFκB-bla THP-1 cell line is responsive to TNFα and can be used to probe NFκB signaling pathways, including those involved in apoptosis, viral defense, cancer, inflammation, and autoimmune disease.
- This cell line can be adapted for high-throughput screening for agonists or antagonists with compound libraries.
- Candidate drugs can be tested for dose response against this cell line.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Product Line | CellSensor |
| Quantity | 1 vial |
| Target Entry | TNF alpha, NIK, TLR, I kappa B, IRAK, IKK, NF-kappa B pathway |
| Type | Cell Line |
| Cell Line | THP-1 |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.