Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF-H Antibody (NAP4), Novus Biologicals™
SDP

Catalog No. NB018133 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018133 0.025 mL
NB300136 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018133 Supplier Novus Biologicals Supplier No. NB3001360.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NF-H Monoclonal specifically detects NF-H in Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen NF-H
Applications Western Blot, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone NAP4
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000
Formulation 50% PBS, 50% Glycerol with 5mM Sodium Azide
Gene Alias 200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H
Gene Symbols NEFH
Host Species Mouse
Immunogen Native NF-H purified from bovine spinal cord
Molecular Weight of Antigen 200-220 kDa
Purification Method Affinity purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4744
Test Specificity Specifically recognizes the phosphorylated variant of NF-H subunit (∼200-220 kDa), showing some weaker reactivity with phosphorylated forms of NF-M.
Target Species Pig, Mouse, Chicken, Rat, Human, Equine, Bovine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.