Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF-H Antibody (NAP4), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | NF-H |
---|---|
Clone | NAP4 |
Concentration | 1 mg/ml |
Dilution | Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000 |
Applications | Western Blot, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry |
Description
NF-H Monoclonal specifically detects NF-H in Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NF-H | |
1 mg/ml | |
Western Blot, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Mouse | |
Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience | |
200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H | |
NEFH | |
IgG1 | |
Affinity purified | |
Specifically recognizes the phosphorylated variant of NF-H subunit (∼200-220 kDa), showing some weaker reactivity with phosphorylated forms of NF-M. |
NAP4 | |
Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000 | |
Monoclonal | |
Ascites | |
RUO | |
Pig, Mouse, Chicken, Rat, Human, Equine, Bovine | |
4744 | |
Native NF-H purified from bovine spinal cord | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
200-220 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title