Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF-H Antibody (NEFL.H/2324R), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB379916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379916 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379916 Supplier Novus Biologicals Supplier No. NBP308552AF405
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NF-H Monoclonal antibody specifically detects NF-H in Human, Mouse, Rat, Porcine, Chicken samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)

Specifications

Antigen NF-H
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone NEFL.H/2324R
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias 200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H
Host Species Rabbit
Immunogen Recombinant full-length human NF-H protein (Uniprot: P12036)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4744
Target Species Human, Mouse, Rat, Pig, Chicken
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.