Learn More
Description
Specifications
Specifications
| Antigen | NF-H |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | NF421 |
| Conjugate | DyLight 680 |
| Dilution | ELISA |
| Gene Alias | 200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H |
| Gene Symbols | NEFH |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human NF-H protein (Uniprot: P12036) |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
