Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF-L Antibody, Novus Biologicals™
SDP

Catalog No. NB300131 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300131 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300131 Supplier Novus Biologicals Supplier No. NB300131
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NF-L Polyclonal antibody specifically detects NF-L in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay.

Specifications

Antigen NF-L
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:10000-1:15000, Immunohistochemistry 1:5000, Immunocytochemistry/Immunofluorescence 1:500-1:5000, Immunohistochemistry-Paraffin 1:5000, Knockout Validated
Formulation Supplied as serum with 5mM Sodium Azide
Gene Accession No. P02547
Gene Alias 68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa
Gene Symbols NEFL
Host Species Rabbit
Immunogen Native Neurofilament Light protein purified from pig spinal cord [UniProt# P02547]
Molecular Weight of Antigen 68 kDa
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4747
Test Specificity Specifically recognizes the light neurofilament subunit (∼68 kDa).
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.