Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF-M Antibody (NF-09), DyLight 680
SDP

Catalog No. NB500446FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500446FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500446FR Supplier Novus Biologicals Supplier No. NB500446FR

Mouse Monoclonal Antibody

NF-M Monoclonal specifically detects NF-M in Mouse, Porcine, Mammal samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen NF-M
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone NF-09
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Alias NEF3neurofilament medium polypeptide, Neurofilament 3, neurofilament, medium polypeptide, neurofilament, medium polypeptide 150kDa, neurofilament-3 (150 kD medium), NF-M160 kDa neurofilament protein, NFMNeurofilament triplet M protein
Gene Symbols NEFM
Host Species Mouse
Immunogen Pellet of pig brain cold stable proteins after depolymerization of microtubules.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cellular Markers, Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4741
Test Specificity The antibody NF-09 reacts with both phosphorylated and non-phosphorylated form of medium neurofilament protein (160 kDa) of various species.
Target Species Mouse, Pig, Mammalia
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.