Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF-M Antibody (OTI2G3), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP272977A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272977A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272977A94 Supplier Novus Biologicals Supplier No. NBP272977AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NF-M Monoclonal specifically detects NF-M in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NF-M
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2G3
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias NEF3neurofilament medium polypeptide, Neurofilament 3, neurofilament, medium polypeptide, neurofilament, medium polypeptide 150kDa, neurofilament-3 (150 kD medium), NF-M160 kDa neurofilament protein, NFMNeurofilament triplet M protein
Gene Symbols NEFM
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 413-773 of human NF-M (NP_005373) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4741
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.