Learn More
NF-YA Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | NF-YA |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2334
![]() |
Bethyl Laboratories, Inc
IHC00576T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2333
![]() |
Bethyl Laboratories, Inc
IHC00576 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Monkey, Gorilla,
The protein encoded by this gene is one subunit of a trimeric complex, forming a highly conserved transcription factor that binds to CCAAT motifs in the promoter regions in a variety of genes. Subunit A associates with a tight dimer composed of the B and C subunits, resulting in a trimer that binds to DNA with high specificity and affinity. The sequence specific interactions of the complex are made by the A subunit, suggesting a role as the regulatory subunit. In addition, there is evidence of post-transcriptional regulation in this gene product, either by protein degradation or control of translation. Further regulation is represented by alternative splicing in the glutamine-rich activation domain, with clear tissue-specific preferences for the two isoforms.Specifications
NF-YA | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
CAAT box DNA-binding protein subunit A, CAAT-box DNA binding protein subunit A, CBF-A, CBF-B, CCAAT-binding transcription factor subunit B, HAP2, HAP2 CCAAT-binding protein, NF-YA, nuclear transcription factor Y subunit A, nuclear transcription factor Y, alpha, Transcription factor NF-Y, A subunit | |
Between 333 and 347 | |
Primary | |
4° C | |
NFYA |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P23511, P23708 | |
18044, 4800 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.