Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFAT5 Antibody, Novus Biologicals™
SDP

Catalog No. NB1203446 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1203446 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1203446 Supplier Novus Biologicals Supplier No. NB1203446

Rabbit Polyclonal Antibody has been used in 8 publications

NFAT5 Polyclonal specifically detects NFAT5 in Human, Mouse, Rat, Canine, Hamster, Rabbit samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Super Shift Assays, Knockdown Validated.

Specifications

Antigen NFAT5
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 3 ug, Immunohistochemistry-Paraffin 1:20, Gel Super Shift Assays 1:1 - 1:100, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockdown Validated
Formulation PBS containing 1 mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. O94916
Gene Alias EC 3.6.1, EC 3.6.3.14, KIAA0827glutamine rich protein H65, NF-AT5NFAT-like protein 1, NFATL1, NFATZ, nuclear factor of activated T-cells 5, tonicity-responsive, OREBP, osmotic response element-binding protein, T-cell transcription factor NFAT5, TonE-binding protein, TonEBP, TONEBPnuclear factor of activated T-cells 5, Tonicity-responsive enhancer-binding protein
Gene Symbols NFAT5
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues C D(1439) L L V S L Q N Q G N N L T G S F(1455) of human NFAT5.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 10725
Test Specificity Mouse reactivity reported in scientific literature (PMID: 19412173). Rat reactivity reported in scientific literature (PMID: 12824075).
Target Species Human, Mouse, Rat, Canine, Hamster, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.