Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFATC1/NFAT2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

Supplier:  Novus Biologicals NB10056732

 View more versions of this product

Catalog No. NB10056732


Only null left
Add to Cart

Description

Description

NFATC1/NFAT2 Polyclonal specifically detects NFATC1/NFAT2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

NFATC1/NFAT2
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
MGC138448, NFAT transcription complex cytosolic component, NFAT2cytoplasmic 1, NFATC, NF-ATC, NFATc1, NF-ATc1, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1
Rabbit
Protein G purified
RUO
Primary
The amino acid sequence used is 100% identical in human NFATc1 A, B, C and E isoforms. The reported amino acid size of NFATc1 isoforms are as follows: A -716 aa, B-825 aa, C-930 aa, and E-812 aa.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunocytochemistry, Immunofluorescence
1.0 mg/mL
Western Blot 1-3 ug/ml, Simple Western 1:100, Immunohistochemistry reported in scientific literature (PMID 35167499), Immunocytochemistry/Immunofluorescence 1:20
NP_006153.2
NFATC1
Amino acids 264-282 (NKRKYSLNGRQPPYSPHHS) of human NFATc1 were used as immunogen for this antibody.
0.1 mg
Wnt Signaling Pathway
4772
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only