Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFATC1/NFAT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$207.00 - $489.50
Specifications
Antigen | NFATC1/NFAT2 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1-3 ug/ml, Simple Western 1:100, Immunohistochemistry reported in scientific literature (PMID 35167499), Immunocytochemistry/Immunofluorescence 1:20 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056732T
![]() |
Novus Biologicals
NB10056732SS |
0.025 mg |
Each for $207.00
|
|
|||||
NB10056732
![]() |
Novus Biologicals
NB10056732 |
0.1 mg |
Each for $489.50
|
|
|||||
Description
NFATC1/NFAT2 Polyclonal specifically detects NFATC1/NFAT2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.Specifications
NFATC1/NFAT2 | |
Western Blot 1-3 ug/ml, Simple Western 1:100, Immunohistochemistry reported in scientific literature (PMID 35167499), Immunocytochemistry/Immunofluorescence 1:20 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
NP_006153.2 | |
4772 | |
Amino acids 264-282 (NKRKYSLNGRQPPYSPHHS) of human NFATc1 were used as immunogen for this antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Wnt Signaling Pathway | |
PBS with 0.05% Sodium Azide | |
MGC138448, NFAT transcription complex cytosolic component, NFAT2cytoplasmic 1, NFATC, NF-ATC, NFATc1, NF-ATc1, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1 | |
NFATC1 | |
IgG | |
Protein G purified | |
The amino acid sequence used is 100% identical in human NFATc1 A, B, C and E isoforms. The reported amino acid size of NFATc1 isoforms are as follows: A -716 aa, B-825 aa, C-930 aa, and E-812 aa. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title