Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFATC1/NFAT2 Rabbit anti-Human, Clone: 4I9P10, Novus Biologicals™
SDP

Catalog No. NB166701 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166701 100 μg
NB166702 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166701 Supplier Novus Biologicals Supplier No. NBP315803100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NFATC1/NFAT2 Monoclonal antibody specifically detects NFATC1/NFAT2 in Human samples. It is validated for Western Blot

Specifications

Antigen NFATC1/NFAT2
Applications Western Blot
Classification Monoclonal
Clone 4I9P10
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias MGC138448, NFAT transcription complex cytosolic component, NFAT2cytoplasmic 1, NFATC, NF-ATC, NFATc1, NF-ATc1, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 844-943 of human NFATC1/NFAT2 (O95644).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4772
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.