Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFATC1/NFAT2 Rabbit anti-Human, Clone: 4I9P10, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | NFATC1/NFAT2 |
---|---|
Clone | 4I9P10 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
NFATC1/NFAT2 Monoclonal antibody specifically detects NFATC1/NFAT2 in Human samples. It is validated for Western BlotSpecifications
NFATC1/NFAT2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Wnt Signaling Pathway | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
4772 | |
IgG | |
Affinity purified |
4I9P10 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
MGC138448, NFAT transcription complex cytosolic component, NFAT2cytoplasmic 1, NFATC, NF-ATC, NFATc1, NF-ATc1, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1 | |
A synthetic peptide corresponding to a sequence within amino acids 844-943 of human NFATC1/NFAT2 (O95644). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title