Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFATC2/NFAT1 Antibody (25A10.D6.D2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB300504 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300504 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300504 Supplier Novus Biologicals Supplier No. NB300504
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 9 publications

NFATC2/NFAT1 Monoclonal specifically detects NFATC2/NFAT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Gel Super Shift Assays, Sandwich ELISA.

Specifications

Antigen NFATC2/NFAT1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 25A10.D6.D2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50 - 1:2000, Immunohistochemistry-Frozen 1:50 - 1:2000, Gel Super Shift Assays 1:1 - 1:100, Sandwich ELISA 1:100 - 1:2000, Chromatin Immunoprecipitation (ChIP) 1-3 ul
Gene Alias NFAT pre-existing subunit, NFAT transcription complex, preexisting component, NFAT1nuclear factor of activated T-cells, cytoplasmic 2, NFATc2, NF-ATc2, NFATp, NF-ATp, NFATPnuclear factor of activated T-cells, preexisting component, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2, preexisting nuclear factor of activated T-cells 2, T cell transcription factor NFAT1, T-cell transcription factor NFAT1
Gene Symbols NFATC2
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues A(51) I S S P S G L A Y P D D V L D Y G L(69) of mouse NFATc2-A, B and C isoforms.
Molecular Weight of Antigen 140 kDa
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Transcription Factors and Regulators, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4773
Test Specificity Detects nuclear factor of activated T-cells (NFAT). This does not cross react with NFAT2 (NFATc, NFATc1).
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.