Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFATC2/NFAT1 Antibody (25A10.D6.D2), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB359429 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359429 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359429 Supplier Novus Biologicals Supplier No. NB300504MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NFATC2/NFAT1 Monoclonal antibody specifically detects NFATC2/NFAT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Gel Super Shift Assays, ELISA, ChIP assay (ChIP)

Specifications

Antigen NFATC2/NFAT1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Gel Supershift, Sandwich ELISA, ChIP Assay
Classification Monoclonal
Clone 25A10.D6.D2
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Gel Super Shift Assays, Sandwich ELISA, Chromatin Immunoprecipitation (ChIP)
Formulation 50mM Sodium Borate
Gene Alias NFAT pre-existing subunit, NFAT transcription complex, preexisting component, NFAT1nuclear factor of activated T-cells, cytoplasmic 2, NFATc2, NF-ATc2, NFATp, NF-ATp, NFATPnuclear factor of activated T-cells, preexisting component, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2, preexisting nuclear factor of activated T-cells 2, T cell transcription factor NFAT1, T-cell transcription factor NFAT1
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues A(51) I S S P S G L A Y P D D V L D Y G L(69) of mouse NFATc2-A, B and C isoforms.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Phospho Specific, Transcription Factors and Regulators, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4773
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.