Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFATC2/NFAT1 Rabbit anti-Human, Clone: JA11-08, Novus Biologicals™
SDP

Catalog No. NBP266974 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266974 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266974 Supplier Novus Biologicals Supplier No. NBP266974

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

NFATC2/NFAT1 Monoclonal specifically detects NFATC2/NFAT1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen NFATC2/NFAT1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JA11-08
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias NFAT pre-existing subunit, NFAT transcription complex, preexisting component, NFAT1nuclear factor of activated T-cells, cytoplasmic 2, NFATc2, NF-ATc2, NFATp, NF-ATp, NFATPnuclear factor of activated T-cells, preexisting component, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2, preexisting nuclear factor of activated T-cells 2, T cell transcription factor NFAT1, T-cell transcription factor NFAT1
Gene Symbols NFATC2
Host Species Rabbit
Immunogen Synthetic peptide within Human NFATC2/NFAT1 aa 21-70/ 925. (SwissProt: Q13469 Human)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Adaptive Immunity, Immunology, Phospho Specific, Transcription Factors and Regulators, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4773
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.