Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFIB Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB290583 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290583 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290583 Supplier Novus Biologicals Supplier No. NBP204121PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NFIB Polyclonal antibody specifically detects NFIB in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen NFIB
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias CCAAT-box-binding transcription factor, CTF, HMGIC/NFIB, NF-I/B, NFI-B, NFIB2, NFIB3, NFI-RED, nuclear factor 1 B-type, Nuclear factor 1/B, nuclear factor I/BNF1-B, TGGCA-binding protein
Host Species Rabbit
Immunogen Maps to a region between residue 225 and 275 of human Nuclear Factor I/B using the numbering given in entry NP_005587.2 (GeneID 4781).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4781
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.