Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFIL3 Antibody (1218A), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB345955 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345955 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345955 Supplier Novus Biologicals Supplier No. FAB8888MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NFIL3 Monoclonal specifically detects NFIL3 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen NFIL3
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 1218A
Conjugate mFluor Violet 500 SE
Dilution Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias E4BP4IL3BP1, Interleukin-3 promoter transcriptional activator, Interleukin-3-binding protein 1, NF-IL3A, NFIL3AE4 promoter-binding protein 4, nuclear factor interleukin 3 regulated protein, nuclear factor interleukin-3-regulated protein, nuclear factor, interleukin 3 regulated, Transcriptional activator NF-IL3A
Host Species Rabbit
Immunogen E. coli-derived recombinant mouse NFIL3/E4BP4, Lys140-Arg462, Accession # O08750
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4783
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.