Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFIL3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP141077 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141077 Supplier Novus Biologicals Supplier No. NBP141077
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NFIL3 Polyclonal specifically detects NFIL3 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen NFIL3
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Immunoprecipitation 2-5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Alias E4BP4IL3BP1, Interleukin-3 promoter transcriptional activator, Interleukin-3-binding protein 1, NF-IL3A, NFIL3AE4 promoter-binding protein 4, nuclear factor interleukin 3 regulated protein, nuclear factor interleukin-3-regulated protein, nuclear factor, interleukin 3 regulated, Transcriptional activator NF-IL3A
Gene Symbols NFIL3
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 412 and 462 of human E4 promoter binding-protein 4 using the numbering given in entry NP_005375.2 (GeneID 4783).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4783
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.