Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFkB p100/p52 Antibody, Novus Biologicals™
SDP

Catalog No. NB10082063 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10082063 0.1 mL
NB017863 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10082063 Supplier Novus Biologicals Supplier No. NB10082063
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NFkB p100/p52 Polyclonal specifically detects NFkB p100/p52 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen NFkB p100/p52
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100∼1:200, Immunohistochemistry-Paraffin 1:50-1:100, Immunofluorescence 1:100-1:200
Formulation PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide
Gene Accession No. Q00653
Gene Alias DNA-binding factor KBF2, H2TF1, Lymphocyte translocation chromosome 10 protein, Lyt10, LYT10LYT-10, nuclear factor NF-kappa-B p100 subunit, nuclear factor of kappa light chain gene enhancer in B-cells 2, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2, nuclear factor of kappa light polypeptide gene enhancer in B-cells 2 (p49/p100), Oncogene Lyt-10, p52
Gene Symbols NFKB2
Host Species Rabbit
Immunogen The antiserum was produced against synthesized non-phosphopeptide derived from human NFkB p100/p52 around amino acids 870∼874 (S-Q-S-V-E).
Molecular Weight of Antigen 120 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4791
Test Specificity Detects endogenous levels of total NFkB p100/p52 protein.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.